missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, HPLC Grade (Far UV), 99.8+%

Catalog No. AA42880K7
Change view
Click to view available options
Quantity:
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42880K7 4 L
AA42880K2 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42880K7 Supplier Thermo Scientific Chemicals Supplier No. 042880K7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable For Far UV Analysis | CAS: 75-05-8 | C2H3N | 41.053 g/mol

It is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C to -45°C
Color Colorless
Density 0.786 g/cm3 at 20°C
Quantity 4 L
Flash Point 5°C (41°F)
Ignition Point 525°C (977°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range >99.8%
Vapor Pressure 73mm Hg at 20°C
UN Number 1648; Colorless
Beilstein 741857
Solubility Information Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons.
Boiling Point 80°C to 82°C
Formula Weight 41.05
Grade HPLC
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 200-835-2
RTECSNumber AL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only