missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, HPLC, MilliporeSigma™

Catalog No. MAX01451
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01451 4 L Glass Bottle
MAX0145P1 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAX01451 Supplier Technidata Supplier No. AX01451
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Absorbance 0.005AU max. at 254nm, 0.01AU max., 0.05AU max. at 220nm, 1.00AU max. at 190nm
Color Colorless
Appearance Clear liquid, free from particulates
Density 0.787
Quantity 4 L
Ignition Point 524°C (975.2°F)
Infrared Spectrum Conforms to standard
Odor Aromatic
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 1ppm max.
Assay Percent Range 99.8% min.
Titratable Acid 0.008meq/g max.
Linear Formula CHCH3CN
Vapor Pressure 9.7 kPa (72.8mm Hg) at 20°C
Formula Weight 41.5
Grade ACS/HPLC/NF/USP
Chemical Name or Material Acetonitrile
Show More Show Less
DOTInformation DOT Class 3, II
Recommended Storage Room Temperature