missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents

Catalog No. BP11704
Change view
Click to view available options
Quantity:
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP11704 4 L Glass Bottle
BP1170N119 19 L NOWPak™ I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP11704 Supplier Fisher BioReagents Supplier No. FLBP1170 4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetonitrile Anhydrous is commonly used in DNA or peptide synthesis.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 1 max. at 190nm, 0.15 max. at 195nm, 0.07 max. at 200nm, 0.05 max. at 205nm, 0.04 max. at 210nm, 0.02 max. at 220nm, 0.001 max. at 254nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Residue after Evaporation 1.0ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Viscosity 0.36 cP at 20°C
UN Number 1648
Water 10ppm max.
Boiling Point 81°C
Formula Weight 41.05
Percent Purity ≥99.9%
Grade DNA Synthesis
Chemical Name or Material Acetonitrile
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT