missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% Formic Acid, LC-MS Ultra, Solstice CHROMASOLV™
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Specifications
Specifications
| Quantity | 2 L |
| Flash Point | 5°C (41°F) |
| Packaging | Glass Bottle |
| UN Number | UN1648 |
| Grade | LC-MS Ultra CHROMASOLV |
| Chemical Name or Material | Acetonitrile With 0.1% Formic Acid |
Safety and Handling
| Hazard Statement | P210-P261-P302 + P352 + P312-P304 + P340 + P312-P337 + P313-P403 + P235 |
| Precautionary Statement | H225-H302 + H312 + H332-H319 |
| DOTInformation | 3/PGII |
| EINECSNumber | 200-835-2 |