missing translation for 'onlineSavingsMsg'
Learn More

Acetone, HPLC Grade, Ricca Chemical

Catalog No. RSOA00114C
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOA00114C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOA00114C Supplier Ricca Chemical Company Supplier No. RSOA00114C
Only null left

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Quantity 4 L
Packaging Amber Glass Bottle
Boiling Point 56°C
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Acetone