missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, HPLC Grade, Ricca Chemical
Specifications
Specifications
| CAS | 67-64-1 |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C3H6O |
| MDL Number | MFCD00008765 |
| Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
| InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
| SMILES | CC(C)=O |
| IUPAC Name | propan-2-one |
| Molecular Weight (g/mol) | 58.08 |
| Show More |