missing translation for 'onlineSavingsMsg'
Learn More

Acetone, for gas chromatography MS SupraSolv™, MilliporeSigma™

Catalog No. M1006581000
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1006581000 1 L Glass Bottle
M1006582500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1006581000 Supplier Technidata Supplier No. 1.00658.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography MS SupraSolv™

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 5 to 6 (at 395g/L 20°C)
Melting Point -95.4°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 1 L
Flash Point <−20°C (<−4°F)
Ignition Point 465°C (869°F)
Odor Fruity
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.35
Vapor Pressure 233 hPa (20°C)
UN Number 1090
Boiling Point 56.2°C
Solubility Information Soluble in water at 20°C
Formula Weight 58.08
Chemical Name or Material Acetone
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H336 May cause drowsiness and dizziness.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.