missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023037
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023037 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023037 Supplier Merck Emd Millipore Supplier No. RTC00007820ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Vapor Pressure 184 mmHg (20°C)
Vapor Density 2 (vs. air)
Density 0.791 g/mL (at 25°C (literature))
Flash Point -17°C (Not Applicable)
Molecular Formula C3H6O
Linear Formula CH3COCH3
MDL Number MFCD00008765
Melting Point -94°C (literature)
Boiling Point 56°C (760 mmHg)(literature)
Synonym Acetone
Quantity 20 mL
Formula Weight 58.08
Grade Certified Reference Material
Show More Show Less