missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetamide, 97%

Catalog No. AC442545000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC442545000 500 g Glass bottle
AC442541000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442545000 Supplier Thermo Scientific Chemicals Supplier No. 442545000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5977-14-0 | C4H7NO2 | 101.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5977-14-0
InChI Key GCPWJFKTWGFEHH-UHFFFAOYSA-N
Synonym acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
PubChem CID 80077
ChEBI CHEBI:28515
IUPAC Name 3-oxobutanamide
SMILES CC(=O)CC(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -53.0°C to +56.0°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H7NO2
Linear Formula CH3C(O)CH2C(O)NH2
Quantity 500 g
Molecular Weight (g/mol) 101.105
Formula Weight 101.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetamide
Show More Show Less

RUO – Research Use Only