missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid R (300 g/L), Ricca Chemical
Supplier: Ricca Chemical Company 13916
Specifications
| Acetic Acid | |
| 7732-18-5,64-19-7 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| acetic acid | |
| 176 | |
| Laboratory | |
| Natural Poly Bottle |
| Reagent | |
| 75.0,22.63 | |
| 78.0,23.56 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 300 g/L | |
| Mixture | |
| CHEBI:15366 | |
| 500 mL | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |