missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid/Periodic Acid Reagent, Reagents

Catalog No. 01336090
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336090 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336090 Supplier Labchem Inc Supplier No. CA0098002.5D1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid/Periodic Acid Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid/Iodine Tetraoxide+N381697
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless to Light Yellow
Packaging Glass Bottle
Specific Gravity 1.0501
Quantity 2.5 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Acetic Acid/Periodic Acid Reagent