missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (USP/FCC), Fisher Chemical

Catalog No. A490212
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A490212 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A490212 Supplier Thermo Fisher Scientific Supplier No. FLA490212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Density 1.048g/cm³
Quantity 2.5 L
Flash Point 40°C
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3COOH
Vapor Pressure 1.52 kPa at 20°C
UN Number 2789
Formula Weight 60.05
Percent Purity 99.5 to 100.5% (USP)
Grade FCC/USP
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation Class 8: Corrosive