missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, Reagent, ACS, 99.7%, Spectrum™ Chemical

Catalog No. p-7156901
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600216 100 mL Amber Glass Bottle
18600227 500 mL Amber Glass Bottle
18600217 500 mL Poly Bottle
18600220 2.5 L Glass Bottle
18600219 2.5 L Poly-Coated Glass Bottle
18600228 6 x 500 mL Amber Glass Bottle
18600218 6 x 500 mL Poly Bottle
18600221 4 L Poly Bottle
18600222 4 L Poly-Coated Glass Bottle
18600224 4 x 2.5 L Glass Bottle
18600223 4 x 2.5 L Poly-Coated Glass Bottle
18600231 6 x 2.5 L Glass Bottle
18600232 6 x 2.5 L Poly-Coated Glass Bottle
18600225 4 x 4 L Poly Bottle
18600226 4 x 4 L Poly-Coated Glass Bottle
18600229 20 L Poly Drum
18600230 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 18600216 Supplier Spectrum Chemical Mfg Cor Supplier No. A1010100ML
Only null left

A1010, 64-19-7, C2H4O2

Spectrum™ Chemical Acetic Acid, Glacial, Reagent, ACS , also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. In its Glacial form (undiluted), it is used as a polar protic solvent. As a solvent it is used for purification by recrystallization of organic compounds and in the production of terephtalic acid. As an ACS grade Reagent, Spectrum Chemical manufactured Acetic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 100%
UN Number UN2789
Percent Purity 99.7%
Grade ACS
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%