missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, ACS Grade, ≥ 99.7%, LabChem™

Catalog No. LC101001
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 2.5 L
Packaging:
Cubitainer™
HDPE Bottle
PVC Coated
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC101001 500 mL Cubitainer™
LC101002 1 L PVC Coated
LC101003 2.5 L HDPE Bottle
LC101003S 2.5 L PVC Coated
LC1010036 6 x 2.5 L HDPE Bottle
LC1010036S 6 x 2.5 L PVC Coated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. LC101001 Supplier LabChem Supplier No. LC101001

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 17°C
Boiling Point 118°C
Color Colorless
CAS Max % 100
Linear Formula CH3COOH
UN Number UN2789
Solubility Information Soluble in water
Formula Weight 60.05
Percent Purity ≥99.7%
Grade ACS
Density 1049kg/m3
Quantity 500 mL
Flash Point 40°C
Odor Vinegar
Packaging Cubitainer™
Physical Form Liquid
Residue after Evaporation ≤0.0010%
Specific Gravity 1g/mL
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/spark/open flames/hot surfaces.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state, and federal regulations.
EINECSNumber 607-002-00-6
Recommended Storage Room Temperature