missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 97+%, ACS reagent

Catalog No. AC423275000
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423275000 500 g Plastic bottle
AC423270250 25 g Plastic bottle
AC423270020 2 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423275000 Supplier Thermo Scientific Chemicals Supplier No. 423275000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114°C
Color White
CAS Min % 97.0
pH 6.7 to 7.3 (5% soln. at 25°C)
CAS Max % 100.0
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 97+%
Linear Formula CH3CO2NH4
Beilstein 02, 96
Flash Point 136°C
Fieser 01,38; 07,11
Merck Index 15, 492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol,slightly soluble in acetone,in methanol: 7.89g/100mL (15°C)
Formula Weight 77.08
Percent Purity ≥97%
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Plastic bottle
Residue after Ignition 0.01% max
Density 1.07
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 211-162-9
RTECSNumber AF3675000
TSCA TSCA

RUO – Research Use Only