missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide 98.0+%, TCI America™

Catalog No. A259825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A259825G 25 g
A2598100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A259825G Supplier TCI America Supplier No. A259825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-66-5
Molecular Formula C4H6N4O3S2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00003105
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
Synonym acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
PubChem CID 1986
ChEBI CHEBI:27690
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O

Specifications

Melting Point 259°C
Color White
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetazolamide
RTECSNumber AC8225000
TSCA Yes