missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer, pH 6.0, EP Buffer Solution 4002200, Ricca Chemical
Ep Buffer Solution 4002200
Supplier: Ricca Chemical Company RI6432
Specifications
| Acetate Buffer | |
| 81.0,16.23,0.7 | |
| 84.0,16.89,0.73 | |
| 1X | |
| 6 | |
| 1 L | |
| Natural Poly Bottle |
| pH 6.0 | |
| Colorless | |
| 64-19-7 | |
| Product Monographs | |
| Liquid | |
| Buffer | |
| Laboratory |