missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagents

Catalog No. 01335948
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
20 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335948 120 mL
01335947 1 L
01335946 20 L
01335945 250 mL
01335944 4 L
01335943 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01335948 Supplier Labchem Inc Supplier No. CA102300120A
Only null left

Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetate Buffer, pH 3.5 For USP Heavy Metals Test
CAS 7732-18-5, 631-61-8, 631-61-8
Boiling Point 118°C
Color Colorless
pH 3.5
Physical Form Liquid
Quantity 120 mL
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.0737
Sterility Non-sterile
Grade Reagent
Show More Show Less