missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014898
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014898 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014898 Supplier Merck Emd Millipore Supplier No. PHR1531200MG

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)

Specifications

CAS 614-80-2
Linear Formula CH3 CONHC6H4 OH
MDL Number MFCD00002181
Melting Point 205°C to 210°C
Synonym 2-Acetamidophenol; 2-Acetamidophenol; 2 ′-Hydroxyacetanilide; N-(2-Hydroxyphenyl)acetamide; NSC 3989
Type Certified Reference Material
Quantity 200 mg
Formula Weight 151.16
Grade Certified Reference Material