missing translation for 'onlineSavingsMsg'
Learn More

Acetamidine Hydrochloride 95.0+%, TCI America™

Catalog No. A0008500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0008500G 500 g
A000825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0008500G Supplier TCI America Supplier No. A0008500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-42-5
Molecular Formula C2H7ClN2
Molecular Weight (g/mol) 94.542
MDL Number MFCD00013016
InChI Key WCQOBLXWLRDEQA-UHFFFAOYSA-N
Synonym acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
PubChem CID 67170
IUPAC Name ethanimidamide;hydrochloride
SMILES CC(=N)N.Cl

Specifications

Melting Point 168°C
Color White
Quantity 500 g
Formula Weight 94.54
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetamidine Hydrochloride