missing translation for 'onlineSavingsMsg'
Learn More

Acetamidine hydrochloride, 97%

Catalog No. AAA1609136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1609136 500 g
AAA1609114 25 g
AAA1609122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1609136 Supplier Thermo Scientific Chemicals Supplier No. A1609136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 124-42-5 | C2H7ClN2 | 94.542 g/mol

Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

Solubility
Soluble in water.

Notes
It is hygroscopic. Store away from oxidizing agents.

Chemical Identifiers

CAS 124-42-5
Molecular Formula C2H7ClN2
Molecular Weight (g/mol) 94.542
MDL Number MFCD00013016
InChI Key WCQOBLXWLRDEQA-UHFFFAOYSA-N
Synonym acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
PubChem CID 67170
IUPAC Name ethanimidamide;hydrochloride
SMILES CC(=N)N.Cl

Specifications

Melting Point 168°C to 175°C
Quantity 500 g
Beilstein 3591762
Sensitivity Hygroscopic
Merck Index 14,44
Solubility Information Soluble in water.
Formula Weight 94.54
Percent Purity 97%
Chemical Name or Material Acetamidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-700-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only