missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%

Catalog No. AAA1258930
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1258930 250 g
AAA125890B 1000 g
AAA125890I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1258930 Supplier Thermo Scientific Chemicals Supplier No. A1258930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60-35-5 | C2H5NO | 59.068 g/mol

Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00008023
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 76°C to 81°C
Density 1.159
Boiling Point 220°C to 222°C
Quantity 250 g
Beilstein 1071207
Sensitivity Hygroscopic
Merck Index 14,43
Solubility Information Soluble in water.
Formula Weight 59.07
Percent Purity 99%
Chemical Name or Material Acetamide
Show More Show Less
Hazard Category H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 200-473-5
RTECSNumber AB4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only