Learn More
Aceclofenac, 99%
CAS: 89796-99-6 | C16H13Cl2NO4 | 354.183 g/mol
$154.63 - $317.75
Chemical Identifiers
| CAS | 89796-99-6 |
|---|---|
| Molecular Formula | C16H13Cl2NO4 |
| Molecular Weight (g/mol) | 354.183 |
| MDL Number | MFCD00864296 |
| InChI Key | MNIPYSSQXLZQLJ-UHFFFAOYSA-N |
| Synonym | aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish |
| PubChem CID | 71771 |
| ChEBI | CHEBI:31159 |
| IUPAC Name | 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6035506
|
Thermo Scientific Chemicals
J6035506 |
5 g |
Each for $154.63
|
|
|||||
|
AAJ6035509
|
Thermo Scientific Chemicals
J6035509 |
10 g |
Each for $317.75
|
|
|||||
Description
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.Chemical Identifiers
| 89796-99-6 | |
| 354.183 | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 71771 | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| C16H13Cl2NO4 | |
| MFCD00864296 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| CHEBI:31159 | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Specifications
| 149°C to 153°C | |
| White | |
| MFCD00864296 | |
| 14,22 | |
| Soluble in DMSO | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl | |
| 354.183 | |
| 71771 | |
| 354.18 | |
| Powder |
| 89796-99-6 | |
| C16H13Cl2NO4 | |
| UN2811 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid | |
| 5 g | |
| CHEBI:31159 | |
| 99% | |
| Aceclofenac |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
H301-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : CY1577900
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only