Learn More
Aceclofenac, 99%
CAS: 89796-99-6 | C16H13Cl2NO4 | 354.183 g/mol
Supplier: Thermo Scientific Chemicals J6035509
Description
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.Specifications
| 149°C to 153°C | |
| 89796-99-6 | |
| C16H13Cl2NO4 | |
| UN2811 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid | |
| 10 g | |
| CHEBI:31159 | |
| 99% |
| Aceclofenac | |
| White | |
| MFCD00864296 | |
| 14,22 | |
| Soluble in DMSO | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl | |
| 354.183 | |
| 71771 | |
| 354.18 | |
| Powder |
Chemical Identifiers
| 89796-99-6 | |
| 354.183 | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 71771 | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| C16H13Cl2NO4 | |
| MFCD00864296 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| CHEBI:31159 | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
H301-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : CY1577900
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only