accessibility menu, dialog, popup

UserName

CAS RN 89796-99-6

HOOOOHNClCl

CAS RN 89796-99-6

IUPAC Name: 2-[(2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetyl)oxy]acetic acid
Synonyms: aceclofenac aceclofenac (product) icn brand of aceclofenac falcol difucrem aital biofenac gerbin difucrem almirall brand of aceclofenac aceclofenaco farma lepori brand of aceclofenac
Molecular Weight (g/mol): 354.18
Molecular Formula: C16H13Cl2NO4
InChi Key: MNIPYSSQXLZQLJ-UHFFFAOYSA-N
SMILES: OC(=O)COC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)