missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac, 99%

CAS: 89796-99-6 | C16H13Cl2NO4 | 354.183 g/mol

Supplier:  Thermo Scientific Chemicals J6035506

 View more versions of this product

Catalog No. AAJ6035506


Only null left
Explore available promotions

Description

Description

A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.
Specifications

Specifications

149°C to 153°C
89796-99-6
C16H13Cl2NO4
UN2811
aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish
MNIPYSSQXLZQLJ-UHFFFAOYSA-N
2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
5 g
CHEBI:31159
99%
Aceclofenac
White
MFCD00864296
14,22
Soluble in DMSO
C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl
354.183
71771
354.18
Powder

Chemical Identifiers

89796-99-6
354.183
MNIPYSSQXLZQLJ-UHFFFAOYSA-N
71771
2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
C16H13Cl2NO4
MFCD00864296
aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish
CHEBI:31159
C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c

H301-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CY1577900

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only