missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
Description
Comprend
Removable bulb protector
For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical
- 0 to 14pH range
- Porous ceramic junction
- Twist-open/twist-close cap for easy filling
- Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
- Immersion depth, ∽4 in. (10.1cm)
- 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead
Choice of Ag/AgCl Single- or Double-Junction internal reference elements
- Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
- Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
- For aqueous and nonaqueous samples
- Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers
Spécifications
Spécifications
| Closure Type | Twist Open/Twist Close |
| Color | Blue |
| Connector Types | BNC |
| Junction | Single Ceramic |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 23°C to 212°C |
| Type | Combination pH Electrode |
| Filling Solution Type | SP135-500 |
| For Use With (Application) | For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical |
| Afficher plus de résultats |