missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Four Cell Conductivity/ATC Probes, Four-Cell

Catalog No. 13620166
Change view
Click to view available options
Cell Constant:
1 cm-1
10 cm-1
Measurement Ranges:
1 to 200 mS/cm
10 μS to 100 mS/cm
Material:
Epoxy
Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Constant Measurement Ranges Material
13620166 10 cm-1 1 to 200 mS/cm Epoxy
13620163 1 cm-1 10 μS to 100 mS/cm Glass
13620165 1 cm-1 10 μS to 100 mS/cm Epoxy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13620166 Supplier Fisherbrand™ Supplier No. 13 620 166

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.

  • Four-cell type with glass or epoxy bodies
  • Built-in temperature sensors and 8-pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1

Specifications

Cell Constant 10 cm-1
For Use With (Equipment) Fisher Scientific accumet benchtop models XL600, XL500, XL200, and AB200 and legacy models XL60, XL50, XL30, XL20, AR60, AR50, AR30, AR20, and AB30
Type Conductivity/ATC Probe
Connector Types 8-pin
Material Epoxy
Measurement Ranges 1 to 200 mS/cm
Storage Method Immersed in deionized water
Probe 4 Cell Conductivity