missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR440 Dissolved Oxygen Meter Kit

Catalog No. 13636AR440A
Change view
Click to view available options
Sensor Types:
Dissolved Oxygen Sensor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensor Types
13636AR440A Dissolved Oxygen Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13636AR440A Supplier Fisherbrand™ Supplier No. 13636AR440A
Get a pH Buffer Kit with pH/mV Meter Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to deliver precise and reliable electrochemistry testing, the accumet AR440 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in., high-resolution color display with capacitive touchscreen operable with gloves.
  • Water-saturated air, air-saturated water, or custom calibration.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5,000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.

Specifications

Measurement Modes Dissolved Oxygen
Sensor Types Dissolved Oxygen Sensor
Range (Polarographic DO) 0.0 to 600.0 % saturation; 0.01 to 90.0 mg/L
Temperature (Metric) Range -30°C to 130°C
Display Type 7-in. High-resolution Color Graphic LCD
Data Points 5000 with Date and Time Stamp
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
Relative Humidity Range 5 to 85%, non-condensing
Percent Saturation Range Polarographic DO 0.0 to 600.0 % saturation
Channels 1
Type Dissolved Oxygen Meter
Voltage 100/240 V
Show More Show Less