missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard/n1000 ppm NH3, Fisherbrand

Catalog No. ACCU0801500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0801500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0801500 Supplier Ricca Chemical Company Supplier No. ACCU0801500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Natural Poly Bottle
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Ion Selective Electrode
Solubility Miscible with water
Solution Type Calibration Standard
Type Calibration Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Ammonia Standard
Show More Show Less