missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Self-Laminating HMCIS Chemical Identifier Labels

Catalog No. 19804618
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19804618 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804618 Supplier Accuform Signs Supplier No. LZS321EV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These labels are essential for labeling containers that contain hazardous chemicals.

Specifications

Material Vinyl
Quantity 100/Roll
Adhesive Type Acrylic
For Use With (Application) To identify the hazardous chemical
Thickness 4 mil
Width (English) 3 in.
Length (English) 1 in.
Length (Metric) 2.54 cm
Product Type HMCIS Chemical Identifier Label
Color White
Life Span 2 to 4 Years
Show More Show Less