Learn More
Accuform Signs Self-Adhesive Individual Numerals
Use numbers and letters to properly mark and identify objects and areas.
Supplier: Accuform Signs NAK1014
Identifiants chimiques
| Vinyl | |
| Multitude of identification needs in various environments | |
| 176°F | |
| -20°C to 80°C | |
| Individual Number |
| 50/Pk. | |
| 3.5 in. | |
| -20°F | |
| -20°F to 176°F | |
| Black on White |
Spécifications
| Vinyl | |
| Multitude of identification needs in various environments | |
| 176°F | |
| -20°C to 80°C | |
| 4 | |
| Black on White |
| 50/Pk. | |
| 3.5 in. | |
| -20°F | |
| -20°F to 176°F | |
| Individual Number |