missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Accessories for DURAN™ Laboratory Glass Bottles
Spécifications
Spécifications
| Color | White |
| Material | Polypropylene, PTFE |
| Pore Size | 0.2 μm |
| For Use With (Application) | Pressure compensation |
| Type | Syringe Disc Filter |