missing translation for 'onlineSavingsMsg'
Learn More

Promega Access RT-PCR System DFS Item

Access RT-PCR System is designed for the reverse transcription and PCR amplification of a specific target RNA from total RNA or mRNA.

Supplier:  Promega A1250

 View more versions of this product

Catalog No. PR-A1250


Only null left
Explore available promotions

Description

Description

  • Convenient RT-PCR in One Tube
  • All the reagents you need for single-tube reverse transcription and PCR
  • Separate components allow you to control every step of the reaction
Specifications

Specifications

Contains sufficient reagents for 100 reactions, 25 reactions containing Positive Control RNA and Primers
• AMV Reverse Transcriptase, 500 μL
• Tfl DNA Polymerase, 500 U
• AMV/Tfl 5X Reaction Buffer, 1 mL
• MgSO, 1.25 mL
• dNTP Mixture, 100 μL
• 50 μL Positive Control

Store at –30° to –10°C

100 Reactions
100 Reactions
Access RT-PCR system
RT-PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions