Learn More
Promega Access RT-PCR System
Access RT-PCR System is designed for the reverse transcription and PCR amplification of a specific target RNA from total RNA or mRNA.
$314.91 - $4,510.15
| Catalog Number | Mfr. No. | Kit Type | No. of Reactions | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Kit Type | No. of Reactions | Price | Quantity & Availability | ||||
PRA1250
|
Promega
A1250 |
Access RT-PCR system | 100 Reactions |
Each for $1,119.96
|
|
||||
PRA1260
|
Promega
A1260 |
Access RT-PCR introductory system | 20 Reactions |
Each for $314.91
|
|
||||
PRA1280
|
Promega
A1280 |
Access RT-PCR system | 500 Reactions |
Each for $4,510.15
|
|
||||
Description
- Convenient RT-PCR in One Tube
- All the reagents you need for single-tube reverse transcription and PCR
- Separate components allow you to control every step of the reaction
Specifications
| RT-PCR |