missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACADS Polyclonal Antibody

Description
Antibody detects endogenous levels of total ACADS.
Introduces a double bond at position 2 in saturated acyl-CoA's of short chain length, i. e. less than 6 carbon atoms.
Spécifications
Spécifications
| Antigen | ACADS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ACADS |
| Gene Accession No. | P15651, P16219, Q07417 |
| Gene Alias | ACAD3; Acads; acetyl-Coenzyme A dehydrogenase, short chain; acyl-CoA dehydrogenase short chain; acyl-CoA dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, short chain; AI196007; Bcd1; Bcd-1; butyryl-CoA dehydrogenase; EGK_04272; Hdlq8; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; short chain acyl-coenzyme A dehydrogenase; short-chain acyl-CoA dehydrogenase; short-chain specific acyl-CoA dehydrogenase, mitochondrial; unsaturated acyl-CoA reductase |
| Gene Symbols | ACADS |
| Afficher plus de résultats |