missing translation for 'onlineSavingsMsg'
Learn More

Acacia, Total ash <4%

Catalog No. AA36499A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36499A3 2 kg
AA3649936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36499A3 Supplier Thermo Scientific Chemicals Supplier No. 036499A3
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Mixture of polysaccharides and glycoproteins used in the food industry as a stabilizer | CAS: 9000-01-5

It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals. In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9000-01-5
MDL Number MFCD00081264
Synonym Gum Arabic

Specifications

Color White
Density 1.4 g/mL
Mesh Size −100 Mesh
Odor Uncharacteristic
Quantity 2 kg
Merck Index 14,14
Solubility Information Water soluble. Aqueous solution is acidic to litmus.
Physical Form Powder
Chemical Name or Material Acacia, Total ash <4%
Hazard Category H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-519-5
RTECSNumber CE5945000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only