missing translation for 'onlineSavingsMsg'
Learn More

Pig Blue™ Absorbent Mat Pad in Dispenser Box

Catalog No. 18366480
Change view
Click to view available options
Dimensions (L x W):
15 x 19 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18366480 15 x 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18366480 Supplier Pig Blue™ Supplier No. BLU106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Soaks up common industrial liquids and delivers more absorbency per pad.

  • Heavyweight
  • Clean up big, persistent leaks and drips, especially in hard-to-reach areas with these heavyweight absorbency pads.
  • Perforated to allow you to tear off small or large pieces
  • Contains 75% recycled fibers
  • Fire retardant for added safety, flame-tested in accordance with ASTM Standard E84-08
  • Absorbs most common industrial liquids, including oils, coolants, solvents, water and other spills
  • 15 x 19 in.
  • 50 pads per box

Specifications

Material Cellulose
Type Absorbent Pad
Disposable Single-use
Weight Capacity Heavyweight
Dimensions (L x W) 15 x 19 in.