missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, low ROX

Catalog No. AB4323A
Change view
Click to view available options
No. of Reactions:
400 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB4323A 400 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB4323A Supplier Thermo Scientific™ Supplier No. AB4323A
Only null left

Incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with ROX or Fluorescein passive reference dyes

Specifications

Concentration 2X
Content And Storage Contains:
• 1 x 5 mL ABsolute Blue QPCR SYBR Green Low ROX Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 400 x 25 μL reactions.

Store at -20°C
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product Line ABsolute Blue
Product Type Real Time PCR SYBR Master Mix
Quantity 400 reactions
Sufficient For 400 Reactions of 25 μL Each
For Use With (Application) Gene Expression, Genotyping, siRNA Validation
Compatibility qPCR Cyclers
No. of Reactions 400 Reactions
Sample Type DNA, dsDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.