missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit

Catalog No. 89024639
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89024639 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89024639 Supplier Abnova™ Supplier No. KA1357

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPAR-alpha, delta, gamma transcription factor DNA binding activity in nuclear extracts and whole cell lysates.

  • Kit is a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates
  • A 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA)
  • A specific double stranded DNA (dsDNA) sequence containing the PPAR response element is immobilized onto the bottom of wells of a 96-well plate
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C

Specifications

Description PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPAR-alpha, delta, gamma transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Immunoassay Kit Format ELISA (Standard)
Product Type PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit
Sufficient For 5 Reactions
For Use With (Application) Functional Study
Gene ID (Entrez) 5465/5467/5468
Gene Symbol PPARA
Quantity 1 Kit
Storage Requirements Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.