Learn More
Danaher Corp Radiometer FLEX PLUS ABL90 Solution Pack (680 Activities)
Supplier:  Danaher Corp 944157
Encompass
ABL90 FLEX PLUS Solution Pack (680 Activities)
680 activities Solution Pack for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
Numéro de catalogue. NC0114692