missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCF3 Polyclonal Antibody

ABCF3 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA5103637
Description
ABCF3 Polyclonal Antibody for Western Blot, ICC/IF
Spécifications
| ABCF3 | |
| Polyclonal | |
| Unconjugated | |
| ABCF3 | |
| ABCF3; AI326318; ATP binding cassette subfamily F member 3; ATP-binding cassette sub-family F member 3; ATP-binding cassette, subfamily F (GCN20), member 3; ATP-binding cassette, sub-family F (GCN20), member 3; AU016058; BB119416; EST201864 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27406, 287982, 55324 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q66H39, Q8K268, Q9NUQ8 | |
| ABCF3 | |
| A synthesized peptide derived from human ABCF3(Accession Q9NUQ8), corresponding to amino acid residues V379-A429. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |