missing translation for 'onlineSavingsMsg'
Learn More

Abamectin

Catalog No. AAJ6003903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6003903 1 g
AAJ6003906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6003903 Supplier Thermo Scientific Chemicals Supplier No. J6003903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Interacts with GABA receptors to cause chloride channel activation | CAS: 71751-41-2 | C48H72O14

Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

Solubility
Soluble in DMSO

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 71751-41-2
Molecular Formula C48H72O14
MDL Number MFCD01769550
Synonym Avermectin B1

Specifications

Melting Point 150°C to 155°C
Color White to Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,2
Solubility Information Soluble in DMSO
Formula Weight 873.1
Physical Form Powder
Chemical Name or Material Abamectin
Hazard Category H300+H330-H361d-H372
Hazard Statement GHS H Statement
H300-H330-H372-H361
Fatal if swallowed.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
RTECSNumber CL1203000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only