missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ab SpinTrap™ Columns
Description
No filtration or centrifugation of different crude samples necessary for antibody purification.
- Protein G Sepharose™ High Performance in prepacked spin columns
- Binds >1mg human IgG/column
- No filtration or centrifugation of different crude samples necessary for antibody purification
- Classic and crosslink protocols for protein enrichment (immunoprecipitation)
- Elution conditions formatted for both LC-MS and electrophoresis analysis
- Binding capacity >1mg IgG/column (determined with human polyclonal IgG)
- Stable in all commonly used buffers within antibody purification
- Contains polypropylene barrel and polyethylene frits
- Average particle size: 34μm
- Bed volume: 100μL
- Maximum sample volume: 600μL
- Short term (2h) pH stability: 2–9
- 600μL undiluted serum sample
Specifications
Specifications
| Bed Volume | 100 mL |
| Description | Ab SpinTrap columns are prepacked, single-use spin columns for rapid purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. Designed for small-scale purification of multiple samples in parallel. |
| Support Type | 6% Highly Cross-linked Agarose |
| Content And Storage | 4 to 8°C, 20% Ethanol |
| Quantity | 50 x 100 μL |
| For Use With (Application) | Centrifugation |
| Type | Spin Column |
| Sample Volume | 600 μl |
| Binding Capacity | >1 mg IgG/Column |
| Capacity (Metric) | 600 μL |