missing translation for 'onlineSavingsMsg'
Learn More

A-7 hydrochloride

Catalog No. AAJ62011MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62011MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62011MB Supplier Thermo Scientific Chemicals Supplier No. J62011MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79127-24-5 | C20H30Cl2N2O2S | 433.432 g/mol

Calmodulin antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79127-24-5
Molecular Formula C20H30Cl2N2O2S
Molecular Weight (g/mol) 433.432
MDL Number MFCD00209822
InChI Key XDJCAQBTSCRBHS-UHFFFAOYSA-N
Synonym a-7 hydrochloride, n-10-aminodecyl-5-chloro-1-naphthalenesulfonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide hydrochloride, 1-10-aminodec-1-yl sulphamoyl-5-chloronaphthalene, n-10-aminodecyl-5-chloro-1-naphthalenesulphonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide-hydrogen chloride 1/1
PubChem CID 44119113
IUPAC Name N-(10-aminodecyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride
SMILES C1=CC2=C(C=CC=C2Cl)C(=C1)S(=O)(=O)NCCCCCCCCCCN.Cl

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble to 50mM in DMSO
Formula Weight 433.44
Physical Form Solid
Chemical Name or Material A-7 hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only