Learn More
Vector Laboratories VECTOR SG HRP SUBSTRATE
Numéro de catalogue. NC9799788
Encompass
VECTOR SG Substrate Kit, 1 kit, Vector SG substrate kit produces a blue/gray reaction product, This substrate can be used singly or as a second label in peroxidase staining
Numéro de catalogue. NC9799788 Fournisseur Vector Laboratories Code fournisseur SK4700
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
VECTOR SG Substrate Kit, 1 kit, Vector SG substrate kit produces a blue/gray reaction product, This substrate can be used singly or as a second label in peroxidase staining