missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates

Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Features:
- Use for high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Spécifications
Spécifications
| Binding Property | Hydrophobic |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 50 Cases, 10 Pk. |
| Product Line | Microtiter |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 280 μL |
| Afficher plus de résultats |