missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural

Catalog No. 14230240
Change view
Click to view available options
Color:
Blue
Green
Natural
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14230240 Green
14230237 Natural
14230239 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14230240 Supplier Fisherbrand™ Supplier No. 14230240

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fit most thermal cyclers

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint

  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced

Specifications

Format Standard
Array 8 x 12
Color Green
Product Type PCR Microplate
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less