Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates
GREENER_CHOICE
Catalog No. 07000720
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Surface Treatment:
Medium Binding
Microlon 600 High Binding
Untreated
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Surface Treatment
07000720 Non-sterile Microlon 600 High Binding
07000091 Non-sterile Untreated
07000082 Sterile Untreated
07000084 Non-sterile Medium Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07000720 Supplier Greiner Bio-One Supplier No. 675061
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9 mm
Service Plan
No
Certifications
DIN ISO 9001
Warranty
No
Specifications
Barcode Alphanumeric
Format Half-Area
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type ELISA Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 600 High Binding
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney
Show More Show Less