Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates
GREENER_CHOICE
Catalog No. 07000084
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Surface Treatment:
Microlon 600 High Binding
Microlon Medium Binding
Non-treated
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Surface Treatment
07000084 Non-sterile Microlon Medium Binding
07000091 Non-sterile Non-treated
07000720 Non-sterile Microlon 600 High Binding
07000082 Sterile Non-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07000084 Supplier Greiner Bio-One Supplier No. 675001
Only null left
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Specifically tre
Certifications
DIN ISO 9001
Specifications
Format Half-Area
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon Medium Binding
Well Shape Flat Bottom (F-well)
No. per Pack 10
Quantity 40/Cs.
Show More Show Less